2-Chloro-p-benzoquinone 95%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Chlorocyclohexa-2,5-diene-1,4-dione, 2-Chloro-1,4-benzoquinone
331220050 331220250
ACRO331220050EA 74.8 EUR
ACRO331220050 ACRO331220250
2-Chloro-p-benzoquinone 95%
2-Chloro-p-benzoquinone
Formula: C₆H₃ClO₂
MW: 142.54 g/mol
Melting Pt: 52…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00075260
CAS Number: 695-99-8
Order Now
Specification Test Results
Appearance Yellow to brown Crystals
Infrared spectrum Conforms
Melting point 52 to 57 °C
GC ≥94 %