Lithium-tert-butanolate ≥99.9% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
44133.04
44133.09
44133.18
44133.04EA
60.3
EUR
44133.04
44133.09
44133.18
Lithium-tert-butanolate ≥99.9% (metals basis)
Lithium tert-butanolate
Purity: 99.9%
CAS No.: 1907-33-1
Molecular Formula: C4H9LiO
Formula Weight: 80.06
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: White to cream
Odour: Odourless
Melting Point: ca 150° dec.
Beilstein Reference: 3620018
UN No.: 3206
MDL No.: MFCD00050479
Formula:
C₄H₉LiO Melting Pt: ∼ 150 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00050479 CAS Number: 1907-33-1 EINECS: 217-611-5 UN: 3206 ADR: 4.2,II |