Lithium-tert-butanolate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
400173-100G 400173-25G
SIAL400173-100GEA 411 EUR
SIAL400173-100G SIAL400173-25G
Lithium-tert-butanolate, Sigma-Aldrich®
Lithium tert-butanolate
Formula: C₄H₉LiO
MW: 80.06 g/mol
MDL Number: MFCD00050479
CAS Number: 1907-33-1
EINECS: 217-611-5
UN: 3206
ADR: 4.2 II