Ethyl cyanoformate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

E18859-10G E18859-50G
SIALE18859-10GEA 82.1 EUR
SIALE18859-10G SIALE18859-50G
Ethyl cyanoformate, Sigma-Aldrich®
Ethyl cyanoformate
Formula: C₄H₅NO₂
MDL Number: MFCD00001836
CAS Number: 623-49-4
EINECS: 210-797-9
UN: 3275
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR