Methyl cyanoformate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

319813-25G 319813-5G
SIAL319813-25GEA 372 EUR
SIAL319813-25G SIAL319813-5G
Methyl cyanoformate, Sigma-Aldrich®
Methyl cyanoformate
Formula: C₃H₃NO₂
CAS Number: 17640-15-2
EINECS: 241-626-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR