Dextran, M.W. ~ 100,000, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
09184-10G-F 09184-250G-F 09184-50G-F
SIAL09184-10G-FEA 32.2 EUR
SIAL09184-10G-F SIAL09184-250G-F SIAL09184-50G-F
Dextran, M.W. ~ 100,000, Sigma-Aldrich®
Dextran
Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5