Ethyl isobutyrylacetate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIALE33203-10GEA 50 EUR
SIALE33203-10G
Ethyl isobutyrylacetate, Sigma-Aldrich®
Ethyl isobutyrylacetate
Formula: C₈H₁₄O₃
MW: 158,2 g/mol
MDL Number: MFCD00009198
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR