Thiazolidine 98%
Supplier: Thermo Fisher Scientific
Danger
165310010 165310050 165310250
ACRO165310010EA 31.2 EUR
ACRO165310010 ACRO165310050 ACRO165310250
Thiazolidine 98%
Thiazolidine
Formula: C₃H₇NS
MW: 89.16 g/mol
Boiling Pt: 72…75 °C (25 mmHg)
Density: 1.131 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00005211
CAS Number: 504-78-9
EINECS: 208-002-5
UN: 2924
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5525 to 1.5545 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥96.0 %