4-Amino-1-benzylpiperidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Amino-N-benzylpiperidine

M-Clarity™ quality level = MQ200

195812-100G 195812-25G 195812-5G
SIAL195812-100GEA 232 EUR
SIAL195812-100G SIAL195812-25G SIAL195812-5G
4-Amino-1-benzylpiperidine, Sigma-Aldrich®
4-Amino-1-benzylpiperidine
Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD00006504
CAS Number: 50541-93-0
EINECS: 256-620-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR