4-Amino-2,2,6,6-tetramethylpiperidine 98%
Supplier: Thermo Scientific
Danger
B24062.06 B24062.14
B24062.06EA 56.9 EUR
B24062.06 B24062.14
4-Amino-2,2,6,6-tetramethylpiperidine 98%
4-Amino-2,2,6,6-tetramethylpiperidine
Purity: 98% CAS No.: 36768-62-4 Molecular Formula: C9H20N2 Formula Weight: 156.27 Flash Point: 75°(167°F) Melting Point: 16-18° Boiling Point: 188-189° MDL No.: MFCD00005984
Formula: C₉H₂₀N₂
MW: 156.27 g/mol
Boiling Pt: 188…189 °C
Melting Pt: 16…18 °C
Density: 0.912
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005984
CAS Number: 36768-62-4
EINECS: 253-197-2