2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥98% free radical
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
TEMPO, 2,2,6,6-Tetramethyl-1-oxylpiperidine, 2,2,6,6-Tetramethylpiperidine 1-oxyl
A12733.03
A12733.06
A12733.14
A12733.XK
A12733.PF
A12733.A1
A12733.03EA
31
EUR
A12733.03
A12733.06
A12733.14
A12733.XK
A12733.PF
A12733.A1
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥98% free radical
2,2,6,6-Tetramethylpiperidinooxy (TEMPO, free radical)
Purity: 98+%
CAS No.: 2564-83-2
Molecular Formula: C9H18NO
Formula Weight: 156.25
Storage Temperature: 2°C to 8°C
Flash Point: 67°(152°F)
Melting Point: 31-40°
Boiling Point: 193° dec.
Beilstein Reference: 1422418
Merck Reference: 14,9140
UN No.: 3263
MDL No.: MFCD00009599
Formula:
C₉H₁₈NO MW: 156.25 g/mol Boiling Pt: 193 °C (dec.) Melting Pt: 31…40 °C Flash Pt: 67 °C (152 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009599 CAS Number: 2564-83-2 EINECS: 219-888-8 UN: 3263 ADR: 8 III Merck Index: 14,9140 |