2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥99%, purified by sublimation, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: TEMPO, 2,2,6,6-Tetramethyl-1-oxylpiperidine, 2,2,6,6-Tetramethylpiperidine 1-oxyl
M-Clarity™ quality level = MQ200
426369-1G 426369-5G
SIAL426369-1GEA 32.6 EUR
SIAL426369-1G SIAL426369-5G
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥99%, purified by sublimation, Sigma-Aldrich®
2,2,6,6-Tetramethylpiperidinooxy (TEMPO, free radical)
Formula: C₉H₁₈NO
MW: 156.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009599
CAS Number: 2564-83-2
EINECS: 219-888-8
UN: 3263
ADR: 8 II