2,5-Dihydroxy-p-benzoquinone, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,5-Dihydroxy-1,4-benzoquinone
M-Clarity™ quality level = MQ100
195464-100G 195464-25G
SIAL195464-100GEA 458 EUR
SIAL195464-100G SIAL195464-25G
2,5-Dihydroxy-p-benzoquinone, Sigma-Aldrich®
2,5-Dihydroxy-p-benzoquinone
Formula: C₆H₄O₄
MW: 140.1 g/mol
MDL Number: MFCD00001598
CAS Number: 615-94-1
EINECS: 210-454-3