3-Amino-5-methyl-1H-pyrazole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Amino-5-methylpyrazole, 5-Methyl-1H-pyrazol-3-amine

M-Clarity™ quality level = MQ200

340200-100G 340200-25G
SIAL340200-100GEA 100 EUR
SIAL340200-100G SIAL340200-25G
3-Amino-5-methyl-1H-pyrazole, Sigma-Aldrich®
3-Amino-5-methyl-1H-pyrazole
Formula: C₄H₇N₃
MW: 97.12 g/mol
MDL Number: MFCD00075180
CAS Number: 31230-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR