2-Amino-4-methoxy-6-methylpyrimidine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
379468-25G
379468-5G
SIAL379468-25GEA
182
EUR
SIAL379468-25G
SIAL379468-5G
2-Amino-4-methoxy-6-methylpyrimidine, Sigma-Aldrich®
2-Amino-4-methoxy-6-methylpyrimidine
Formula:
C₆H₉N₃O MW: 139.16 g/mol |
MDL Number:
MFCD00059768 CAS Number: 7749-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...