2-(Bromomethyl)-1,3-dioxolane, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Bromomethyl-1,3-dioxolane
M-Clarity™ quality level = MQ200
226122-100G 226122-25G
SIAL226122-100GEA 153 EUR
SIAL226122-100G SIAL226122-25G
2-(Bromomethyl)-1,3-dioxolane, Sigma-Aldrich®
2-(Bromomethyl)-1,3-dioxolane
Formula: C₄H₇BrO₂
MW: 167 g/mol
MDL Number: MFCD00003214
CAS Number: 4360-63-8
EINECS: 224-443-6