α-Ketoglutaric acid monosodium salt, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

K2010-100G K2010-25G K2010-5G
SIALK2010-100GEA 737 EUR
SIALK2010-100G SIALK2010-25G SIALK2010-5G
α-Ketoglutaric acid monosodium salt, Sigma-Aldrich®
α-Ketoglutaric acid monosodium salt
Formula: C₅H₅NaO₅
MDL Number: MFCD00064196
CAS Number: 22202-68-2
EINECS: 244-836-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR