1-Benzyl-1H-pyrazole-4-boronic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: (1-Benzyl-1H-pyrazol-4-yl)boronic acid

720445-250MG 720445-1G
SIAL720445-250MGEA 85.9 EUR
SIAL720445-250MG SIAL720445-1G
1-Benzyl-1H-pyrazole-4-boronic acid, Sigma-Aldrich®
1-Benzyl-1H-pyrazole-4-boronic acid
Formula: C₁₀H₁₁BN₂O₂
MW: 202.02 g/mol
MDL Number: MFCD04038760
CAS Number: 852362-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR