Trimellitic anhydride chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1,3-Dioxo-1,3-dihydroisobenzofuran-5-carbonyl chloride, 1,2,4-Benzenetricarboxylic acid 1,2-anhydride 4-chloride

M-Clarity™ quality level = MQ100

T68020-100G T68020-500G
SIALT68020-100GEA 66.6 EUR
SIALT68020-100G SIALT68020-500G
Trimellitic anhydride chloride, Sigma-Aldrich®
Trimellitic anhydride chloride
Formula: C₉H₃ClO₄
MW: 210.57 g/mol
MDL Number: MFCD00005924
CAS Number: 1204-28-0
EINECS: 214-874-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR