Benzeneseleninic anhydride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Benzeneseleninic acid anhydride

M-Clarity™ quality level = MQ200

213012-1G 213012-5G
SIAL213012-1GEA 37.4 EUR
SIAL213012-1G SIAL213012-5G
Benzeneseleninic anhydride, Sigma-Aldrich®
Benzeneseleninic anhydride
Formula: C₁₂H₁₀O₃Se₂
MW: 360.13 g/mol
MDL Number: MFCD00001991
CAS Number: 17697-12-0
EINECS: 241-701-3
UN: 3283
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR