Thiophene-2-carbonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Thiophenecarbonyl chloride, Thiophene-2-carboxylic acid chloride

M-Clarity™ quality level = MQ200

288985-100G 288985-25G
SIAL288985-100GEA 87 EUR
SIAL288985-100G SIAL288985-25G
Thiophene-2-carbonyl chloride, Sigma-Aldrich®
Thiophene-2-carbonyl chloride
Formula: C₅H₃ClOS
MDL Number: MFCD00005428
CAS Number: 5271-67-0
EINECS: 226-092-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR