(-)-Shikimic acid

Supplier: Roth Carl

Synonyms: (3R,4S,5R)-3,4,5-Trihydroxycyclohex-1-ene-1-carboxylic acid, Shikimic acid

7305.1 7305.2
ROTH7305.1EA 99.6 EUR
ROTH7305.1 ROTH7305.2
(-)-Shikimic acid
(-)-Shikimic acid
Formula: C₇H₁₀O₅
MW: 174.15 g/mol
MDL Number: MFCD00066278
CAS Number: 138-59-0
EINECS: 205-334-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR