2,2,6,6-Tetramethyl-4-piperidone monohydrate
Supplier: Thermo Scientific
Synonyms: Triacetonamine monohydrate, 2,2,6,6-Tetramethyl-4-piperidone Hydrate
AC42061.1 AC42061EA AC42061.50
MAYBAC42061.1EA 0 EUR
MAYBAC42061.1 MAYBAC42061EA MAYBAC42061.50
2,2,6,6-Tetramethyl-4-piperidone monohydrate
2,2,6,6-Tetramethyl-4-piperidone monohydrate
Formula: C₉H₁₇NO·1H₂O
MW: 173.26 g/mol
MDL Number: MFCD00149393
CAS Number: 10581-38-1
EINECS: 212-554-2