2,2,6-Trimethyl-4H-1,3-dioxin-4-one, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,2,6-Trimethyl-1,3-dioxine-4-one
M-Clarity™ quality level = MQ200
691380-180KG 691380-1KG 691380-25KG
SIAL691380-180KGEA 16540 EUR
SIAL691380-180KG SIAL691380-1KG SIAL691380-25KG
2,2,6-Trimethyl-4H-1,3-dioxin-4-one, Sigma-Aldrich®
2,2,6-Trimethyl-4H-1,3-dioxin-4-one
Formula: C₇H₁₀O₃
MW: 142.15 g/mol
MDL Number: MFCD00040468
CAS Number: 5394-63-8
EINECS: 226-403-3