Hand disinfectants, Sterillium®
Supplier: Bode
Sterillium®
BODL006690EA 0 EUR
BODL006690 BODL980001 BODL980006_U BODL980007 BODL106650 BODL9804100 BODL669
Hand disinfectants, Sterillium®
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
The propanol based hand disinfectant is ideal for use in all areas of the health industry (operating theatres) and hygiene sensitive industries.
  • Comprehensively active against bacteria, fungi and enveloped viruses
  • Protects against infections in public facilities and when traveling
  • Moisturising, kind to skin with excellent skin tolerance even with long term use
  • Easy to use, rub-in product with surgical standard disinfection in 90 seconds
Upozornení: Use biocides safely. Always read the label and product information before use.
* These articles are only available in Germany.