Disinfectant Solutions, Helipur H Plus N
Supplier: B. Braun
3891950 3892212
148-4142EA 56.3 EUR
148-4142 148-4144
Disinfectant Solutions, Helipur H Plus N
Cleaning Agents Disinfectants
Concentrated disinfectant for temperature-sensitive instruments, endoscopes, in metal, glass or ceramic. Bactericide (inc. TbB), fungicide and virucide (hepatitis B, HIV retrovirus, etc.). Application concentration: 1,5%/60 min; 2,5%/15 min.
It is advisable to read the package leaflet before using this product.
Upozornení: Helipur H Plus is corrosive.