Fix RAL 555
Supplier: RAL
RAL 555
720-0361EA 49.06 EUR
720-0361 RALA362870-2500_U
Fix RAL 555
Stains Microscopy and staining reagents
Reagent Fix RAL 555 is used in RAL 555 kit, Fast version of May-Grunwald Giemsa Staining.
  • Sensitivity, reading and interpretation of the staining are similar to those of traditional May-Grunwald Giemsa stainings.
  • Ideal for emergency diagnosis thanks to its quick execution.
  • Formulation and packaging have been optimised for the user confort and safety.
Sensitivity, reading and interpretation of the staining kit RAL 555 are identical to those of conventional stains. Speed of execution makes it an ideal color for emergency diagnostics.
Cytology Studies of Medullary and Blood Smears in 15 seconds (May-Grunwald Giemsa standard staining in 14 minutes).
Application - Processing time
Haematology, Blood smears = 15 secondes
Haematology, Bone marrow = 3 minutes
Cyto-bacteriology, Liquid Cytologyc investigation (Needle punctures, urine) = 15 secondes
Parasitology, Plasmodium detection test = 1 minute and 4 secondes
The RAL 555 kit (ref 361550) is very easy to use since it consists in successive baths with direct immersion of the slide in the vials.
• The color of the containers results in an enhanced visibility and in a more rapid identification of the various solutions.
• The vials, with broaders necks, make it easier to insert the slide. The kit packaging must not be discarded as it serves as a test stand.
Components - Name (ref) Packing
FIX RAL 555 (Ref 362870) 100 ml (Other packagings 1000 ml, 2500 ml)
EOSIN RAL 555 (Ref 361640) 100 ml (Other packagings 1000 ml, 2500 ml)
BLUE RAL 555 (Ref 361650) 100 ml (Other packagings 1000 ml, 2500 ml)