VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
391-1261EA 114 EUR
391-1261 391-1262
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Seals Microplate Seals
A 139,7 µm thick hydrophobic porous film with medical grade adhesive for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth. These rayon films minimise cross-contamination, spillage and evaporation. They allow uniform air and CO₂ exchange for all wells, unlike plate lids which favour exchange for wells near plate edges. Sterile product is packed in tamper evident bags of 25.
  • Non-cytotoxic, highly gas permeable
  • Pierceable using pipette tip for sample recovery
  • Recommended for temperatures from −20 to +80 °C
Each film L×D: 142,9×82,6 mm for standard size tissue culture plates.