Microbiological media in ampoules
Supplier: Cytiva (Formerly Pall Lab)
68107 4307 4352 68105 68106 68111 4302-N 4306-N
516-7967EA 127 EUR
516-7967 516-7964 516-9081 516-7965 516-7966 516-0423 516-9076 516-9077
Microbiological media in ampoules
Media for Microbiology Prepared Media for Microbiology
A wide variety of sterile ampoule media for common microbiological analyses, in clear glass or plastic ampoules. Supplied pre-measured for maximum efficiency and convenience. For public wastewater and drinking water: MF-Endo and M-FC (with and without rosolic acid for helping to increase the specificity of the medium for faecal coliforms) media are used when analysing water samples for total coliforms, faecal coliforms and E. coli. For food and beverage: M-TGE, HPC, M-Green YM, and orange serum broths are used in the food and beverage industry to QC final products (and fluids used in the manufacture of these products) to detect potential spoilage organisms. Trypticase soy broth, M-TGE, and HPC may be used to test final products and process waters for total bacteria in the pharmaceutical and microelectronics industries. Pseudomonas and KF-Streptococcal broths are highly selective media for organisms in these genera, and are used to monitor samples wherever contamination by these organisms must be prevented.
  • 2 ml ampoules are a practical, time saving aid for culturing microorganisms on membrane filters
  • Ready to use, no dilutions necessary
  • Shelf life up to 1 year storing at 2 to 8 °C
  • Meet Membrane Filter (MF) Technique Standard Methods requirements with 2 ml ampoules (requirements referenced in Standard Methods for the Examination of Water and Wastewater, 1992 APHA, AWWA, WEF, 20th Edition)