VWR®, Ultra-Clear Films for qPCR
Supplier: VWR Collection
391-1258EA 181 EUR
391-1258 391-1295
VWR®, Ultra-Clear Films for qPCR
Seals Microplate Seals
Transparent polyester films with strong, non absorbing, non fluorescing, medical grade adhesive for superior performance in qPCR applications. Supplied non sterile.
  • Recommended for temperatures from −40 to +120 °C
  • Ultra-high optical clarity
  • Certified free from DNase, RNase and nuclease
Each film L×D: 142,9×79,4 mm
Length with end tabs removed: 121,9 mm.
Length with end-tabs removed: 121,9 mm