VWR®, Aluminium Foil Seals for Cold Storage
Supplier: VWR Collection
391-1275EA 128 EUR
391-1275 391-1296
VWR®, Aluminium Foil Seals for Cold Storage
Seals Microplate Seals
Aluminium foils with biologically inert adhesive. These foils are easily pierced by manual or automated pipette tips, for sample recovery without generation of aerosols or cross-contamination.
  • Recommended for temperatures from −80 to +120 °C
  • Not recommended for PCR
Each foil L×D: 133×82 mm