Microtome blades, Personna Plus®
Supplier: Personna
Personna Plus®
PERS72-0114-0000EA 248 EUR
PERS72-0114-0000
Microtome blades, Personna Plus®
Microtomes and Microtome Assemblies Blades and Handles
Personna Plus® microtome blades with MicroCoat® coating.
  • MicroCoat® coating allows blades to glide with smooth and clean precision
  • Uniform ribbon lacking any grooves or split sections of paraffin
  • Transparent viewing area enabling the user to see the number of blades
  • Disposal reservoir prevents used blades from dropping out
Delivery information: In a plastic safety dispenser.