VWR® Ergonomic High Performance (EHP), Multi-Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
613-5254EA 911 EUR
613-5254 613-5257 613-7142 613-7143 613-7144 613-7145 613-7146 613-7147 613-7148 613-7149
VWR® Ergonomic High Performance (EHP), Multi-Channel Pipettes, Mechanical, Variable Volume
Pipettes Micropipettes
The ergonomic high performance pipettes from VWR® provide accuracy and precision you can rely on. The upgraded ergonomic features deliver comfortable pipetting even during extensive periods of use.
  • Significantly reduced plunger forces
  • Effortless volume adjustment with volume lock
  • Superior accuracy and precision for critical applications
  • Tapered shaft and adjustable ‘Easy Grip’ ejector for universal tip fit
  • Extended finger rest for convenient pipetting
  • Fully autoclavable