Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
910725
910410-WA
910425
910710
911010
662-0780EA
52.7
EUR
662-0780
662-1143
662-0777
662-1144
662-1147
Buffer solutions in sachets, perpHect® Pouches, Orion™
Buffers
pH Calibration Buffers
Solutions in sachets ensure optimum precision. The sachet provides a perfect liquid seal and good protection from light.
- Disposable
- Convenient, colour coded sachet and solution for ease of use
- No danger of contamination
- Traceable to NIST standards
Colour coded: pH 4,01 buffer is pink, pH 7,00 buffer is yellow, and pH 10,01 buffer is blue.
There is no wasted buffer or secondary reservoir. Each sachet contains lot code, expiration date, and buffer temperature table.
The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards.