VWR® Culture Media, Ready To Use, In Plates
Supplier: Avantor
VWR®
171274ZAEA 79.5 EUR
171274ZA 22.16.55.153 106820ZA 101004ZAMP 102310CF 170692ZA 600343GF 101114ZA 100521ZA 100880EA 100582JA 122230CA 100460XF 105452ZA 291370ZA 100253ZF 101320ZA 22.16.90.135 101004ZA 100582ZA 100731ZAMP 100250DF 101611ZA 100613DA 101250ZA 100544ZA 100774ZAMP 100053UAMP 176181ZA 100353OF 102898KAMP 171212ZA 171250ZA 102950IMMP 100772ZA30 100884ZA 101241AF 171274ZAMP 100542YA 100774ZA 100544ZAMP 170774ZA 100460ZF 102470ZF 100070ZAMP 175584ZA 100253ZFMP 100641ZA 170884ZA 100470ZA 171023ZA AX051112 100158JF 100782XA 102310XF 173554ZA 100582XAMP 120070ZA 100613IF AX051105 170082UA 101345ZA 101591ZF 171300ZAMP 170731ZA 170884CF 295025ZA AX051101 104744ZA 101611ZAMP 120462AF M90MEDIUM 170460GF 112321TA 175350ZA 100580ZA 100253NF 106370ZF 131112RY 120774ZA 170560ZAMP 122950IM 173580ZA 120053UA 100101ZA 171320ZA 170793ZAMP AX051132 AX051131 172321ZA 101114IFMP 122898KA 101114IF 100882ZA 101203ZAMP 171141AA 120063UA 200460GF AX051121 170793NA AX051120 AX051124 102918ZA 100462AF 100641ZAMP 121203ZA 100980ZA 100692PA 101610AA 170560ZA 173554ZAMP 171380ZA 170692ZAMP 101112DA 100371ZA 101274ZA 170802ZI 100720ZA 100063UA 171002ZA 103554ZA 22.16.90.556.2 AX051150 121294ZA 100581AA 100611ZA 171142AF 22.16.90.036.2 105406ZF 101294ZAMP 100884CF 101114SA 102898KA 100884ZAMP 120460ZF 103542ZF 122310XF 191114ZT 100731ZA 175566ZA 100582ZAMP 101114AF 100852ZA 101274ZAMP 103580ZF 101023ZA 175592ZA 100582JAMP 121004ZA 171400ZAMP 101938ZF 22.16.90.555.2 100782ZA 170542YA 100222ZA 171400ZA 101241ZF 101203ZA 101011ZA 101114ZAMP 100191ZF AX051165 AX051163 100053UA AX051169 AX051168 AX051166 100793ZA 175160ZA 106822ZAB 101215ZA 121114ZA 100331ZA 100070ZA 170793ZA 102950IM 120582ZA 102322ZA 105190ZA 170460ZF 106140AA 105442AF 105690XF 112230CA 101142AF 120460GF 100560ZAMP 100692ZAMP 100331ZAMP 172324ZA 173241ZA 170641ZA 100882EA 102684XA 11.1285 100692ZA 171114ZA 100082XA 173580ZAMP 102685ZA 103660XF 106050ZA 175452ZA 101294ZA 100793ZAMP 100742ZA 100911ZA 171320ZAMP 111114NF 104852AF 106181ZA 101114AFMP 101183ZA 106060ZA 102502ZA 170540YA 102770ZA 103332YA 175880ZA 100460GF 175160ZAMP 120884CF 100782ZAMP 100611ZAMP 100063UAMP 106170ZA 106340ZA 100542CA 101141AA 100121ZA 100560ZA 100582XA 103650YF 170980ZA 105906YF 103662ZA 100793NA 171300ZA 100544XA
VWR® Culture Media, Ready To Use, In Plates
Media for Microbiology Prepared Media for Microbiology
For various microbiological applications.
  • Packed under cleanroom conditions to avoid contamination
  • Manufactured with formulations that follow specifications of the various Pharmacopoeias and ISO regulations
  • Special packaging for 55 mm plates
The 55 mm plate's special packaging has several advantages: Good plate protection during transport and storage, can be reused to transport the plates after use, and is recyclable (PET). The plates can be stored from 2 to 14 °C.
Certifikace: *Chromocult® is a registered trademark of Merck KGaA, Darmstadt, Germany. This ready to use medium is made using only Chromocult® dehydrated media produced by Merck KGaA and Agar.
Balení: Cellophane bag packaging containing 3×10 or 3×3×10 (55 mm plates) or 4×5 or 6×4×5 (90 mm plates).