VWR® Contact Plates, Blister
Supplier: VWR Chemicals
The range is manufactured from high quality raw materials. All culture media are manufactured using selected dehydrated bases, and comply with the requirements of ISO 11133 (food microbiology). Manufactured under GMP, the media is produced using the latest technology and is subject to strict quality controls carried out by qualified microbiologists.
- Packed under cleanroom conditions to avoid contamination
- Storage at 2 to 8 °C
- Manufactured under GMP with formulations that follow specifications of the various Pharmacopoeias and ISO regulations