Sporous Glass Beads
Supplier: ROBU GLASFILTER
80068 80080 80085
ROBU80068EA 596 EUR
ROBU80068 ROBU80080 ROBU80085
Sporous Glass Beads
Beads Glass Beads
Substrate for cell cultivation.
  • Defined pore structure supports uniform cell distribution and growth
  • Inert and biocompatible - ideal for sensitive cell cultures
  • Reusable: Withstands repeated autoclaving and cleaning
  • Available in 3 bead sizes: 1,5, 4,0 and 8,0 mm
  • High thermal resistance up to 500 °C
  • Suitable for research, biotech, and clinical applications
  • Made from high-purity, binder-free borosilicate glass 3.3
VitraPOR® microcarriers are premium-quality beads made of high-purity, binder-free borosilicate glass 3.3, specifically designed to support the growth and expansion of adherent cells. Their defined porous structure provides a highly biocompatible and chemically inert surface, ensuring safe and stable conditions for various cell culture applications.
The open-pore architecture facilitates even cell distribution and minimises cell clumping by allowing cells to attach both on the surface and within the inner pore structure. This promotes uniform growth and enables higher cell density, ideal for applications requiring robust and consistent proliferation - such as in biotechnology, pharmaceutical development, and preclinical research.
*Also available as PU 250 ml on request.