1,3-Dioxane ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: m-Dioxane, 1,3-Dioxacyclohexane
H54107.06 H54107.14
H54107.06EA 68.1 EUR
H54107.06 H54107.14
1,3-Dioxane ≥98%
1,3-Dioxane
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 105…106 °C
Melting Pt: –45 °C
Density: 1.032
Flash Pt: 5 °C (41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006566
CAS Number: 505-22-6
EINECS: 208-005-1
UN: 1165
ADR: 3 II