Barium chloride 20% in aqueous solution
Supplier: VWR Chemicals
240314.5000EA 129 EUR
240314.5000
Barium chloride 20% in aqueous solution
Barium chloride
Formula: BaCl₂
MW: 208,24 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
MDL Number: MFCD00003445
UN: 3287
ADR: 6.1,III