2-Bromo-9-fluorenone 96%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Bromo-9H-fluoren-9-one
H55962.MC H55962.ME H55962.14
H55962.MCEA 0 EUR
H55962.MC H55962.ME H55962.14
2-Bromo-9-fluorenone 96%
2-Bromo-9-fluorenone
Formula: C₁₃H₇BrO
MW: 259.1 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00010789
CAS Number: 3096-56-8