2,4-Dichloro-5-methylpyrimidine ≥98%
Supplier: Thermo Fisher Scientific
Danger
H55206.03 H55206.06
H55206.03EA 70.9 EUR
H55206.03 H55206.06
2,4-Dichloro-5-methylpyrimidine ≥98%
2,4-Dichloro-5-methylpyrimidine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 108…109 °C (11 mmHg)
Melting Pt: 26…28 °C
Density: 1.39
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00023197
CAS Number: 1780-31-0
EINECS: 217-227-8
UN: 3261
ADR: 8 II