9-Aminophenanthrene 96%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Phenanthren-9-amine
H55773.MD H55773.03
H55773.MDEA 226 EUR
H55773.MD H55773.03
9-Aminophenanthrene 96%
9-Aminophenanthrene
Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Boiling Pt: 408
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00001177
CAS Number: 947-73-9
EINECS: 213-431-6