4-Aminohippuric acid
Supplier: Roth Carl
Synonyms: p-Aminohippuric acid, 4-Aminohippuric Acid, 2-(4-Aminobenzamido)acetic acid, N-(4-Aminobenzoyl)glycine
6187.1 6187.2
ROTH6187.1EA 0 EUR
ROTH6187.1 ROTH6187.2
4-Aminohippuric acid
4-Aminohippuric acid
Formula: H₂NC₆H₄C(O)NHCH₂COOH
MW: 194.19 g/mol
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 199…202 °C
Density: 1.402 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007890
CAS Number: 61-78-9
EINECS: 200-518-9
Merck Index: 13,00441