1-Acetyl-2-naphthol
Supplier: Thermo Scientific
Synonyms:
1-Acetyl-2-hydroxynaphthalene, 2-Hydroxy-1-acetonaphthone, 1-(2-Hydroxynaphthalen-1-yl)ethanone, 2'-Hydroxy-1'-acetonaphthone, 1-(2-Hydroxy-1-naphthyl)ethan-1-one
SB01921.1
SB01921EA
SB01921.50
MAYBSB01921.1EA
0
EUR
MAYBSB01921.1
MAYBSB01921EA
MAYBSB01921.50
1-Acetyl-2-naphthol
1-Acetyl-2-naphthol
Formula:
C₁₂H₁₀O₂ MW: 186.21 g/mol Boiling Pt: 178 °C (13 mmHg) Melting Pt: 64 °C |
MDL Number:
MFCD00012131 CAS Number: 574-19-6 EINECS: 209-369-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...