Octyl isocyanate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
329746-25G 329746-5G
SIAL329746-25GEA 209 EUR
SIAL329746-25G SIAL329746-5G
Octyl isocyanate, Sigma-Aldrich®
Octyl isocyanate
Formula: C₉H₁₇NO
Boiling Pt: 78 °C (1013 hPa)
Density: 0.88 g/cm³ (20 °C)
MDL Number: MFCD00013887
CAS Number: 3158-26-7
EINECS: 221-598-1
UN: 2206
ADR: 6.1,III