trans-3-Heptene 97
Supplier: Thermo Fisher Scientific
Danger
Synonyms: (E)-3-Heptene
H53482.03 H53482.06 H53482.14
H53482.03EA 66.9 EUR
H53482.03 H53482.06 H53482.14
trans-3-Heptene 97
trans-3-Heptene
Formula: C₇H₁₄
MW: 98.19 g/mol
Boiling Pt: 94 °C
Melting Pt: –137 °C
Density: 0.698
Flash Pt: –1 °C (30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009475
CAS Number: 14686-14-7
EINECS: 238-728-8
UN: 3295
ADR: 3 II