6-Bromoveratraldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: 6-Bromo-3,4-dimethoxybenzaldehyde, 2-Bromo-4,5-dimethoxybenzaldehyde

M-Clarity™ quality level = MQ100

155578-25G 155578-5G
SIAL155578-25GEA 414 EUR
SIAL155578-25G SIAL155578-5G
6-Bromoveratraldehyde, Sigma-Aldrich®
6-Bromoveratraldehyde
Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 151 °C
MDL Number: MFCD00003301
CAS Number: 5392-10-9
EINECS: 226-390-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR