Titanium(IV) chloride 1 M in dichloromethane
Supplier: Thermo Scientific
Danger
Synonyms: Titanium tetrachloride
H31830.AE H31830.AP
H31830.AEEA 59 EUR
H31830.AE H31830.AP
Titanium(IV) chloride 1 M in dichloromethane
Titanium (IV) chloride
CAS No.: 7550-45-0 Molecular Formula: Cl4Ti Formula Weight: 189.69 UN No.: 2922 MDL No.: MFCD00011267
Formula: Cl₄Ti
MW: 189.69 g/mol
Density: 1.365
Storage Temperature: Ambient
MDL Number: MFCD00011267
CAS Number: 7550-45-0
EINECS: 231-441-9
UN: 2922
ADR: 8,III