Dichloro(chloromethyl)methylsilane 95%
Supplier: Thermo Scientific
|
Danger
|
313370250
313371000
ACRO313370250EA
32.9
EUR
ACRO313370250
ACRO313371000
Dichloro(chloromethyl)methylsilane 95%
Dichloro(chloromethyl)methylsilane
Formula:
C₂H₅Cl₃Si MW: 163.51 g/mol Boiling Pt: 121…122 °C Density: 1.26 g/cm³ Flash Pt: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000874 CAS Number: 1558-33-4 EINECS: 216-319-5 UN: 2986 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to straw yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4480 to 1.4510 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...