Gold(III) acetate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
39742.02 39742.03 39742.06
39742.02EA 171 EUR
39742.02 39742.03 39742.06
Gold(III) acetate ≥99.9% (metals basis)
Gold (III) acetate
Purity: 99.9% CAS No.: 15804-32-7 Molecular Formula: Au(O2CCH3)3 Formula Weight: 374.10 Physical Form: Powder
Formula: C₆H₉AuO₆
MW: 374.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210614
CAS Number: 15804-32-7
EINECS: 244-137-6